Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281425.1 | NP_001268354.1 | 2 | 225 | ||
NM_001281426.1 | NP_001268355.1 | 2 | 225 | |||
NM_001281427.1 | NP_001268356.1 | 2 | 225 | |||
NM_007234.4 | NP_009165.1 | 2 | 225 | |||
NM_024348.3 | NP_077324.1 | 2 | 225 | |||
GenBank mRNA | AF082513.1 | - | 2 | 182 | ||
AK091661.1 | - | 2 | 220 | |||
AK314730.1 | - | 2 | 193 | |||
AW953496.1 | - | 2 | 170 | |||
BC000319.2 | - | 2 | 176 | 2821720 | ||
BC003004.1 | - | 2 | 176 | 2821720 | ||
BC107697.1 | - | 2 | 194 | 6454606 | ||
BG715167.1 | - | 2 | 206 | |||
BI596981.1 | - | 2 | 178 | |||
BI837796.1 | - | 2 | 184 | |||
CR541889.1 | - | 2 | 166 | |||
DA212054.1 | - | 2 | 231 |