Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172064.1 | NP_742061.1 | 3 | 126 | ||
XM_006237455.1 | XP_006237517.1 | 3 | 152 | |||
GenBank mRNA | K02247.1 | - | 3 | 164 | ||
L17317.1 | - | 3 | 126 |