Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elavl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172324.1 | NP_758827.1 | 3 | 283 | ||
XM_006242616.2 | XP_006242678.1 | 3 | 627 | |||
XM_008765920.1 | XP_008764142.1 | 3 | 470 | |||
GenBank mRNA | AB097198.1 | - | 3 | 283 |