Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172331.1 | NP_758834.1 | 2 | 104 | ||
XM_006236631.2 | XP_006236693.1 | 2 | 213 | |||
GenBank mRNA | AY150169.1 | - | 2 | 104 | ||
FQ215758.1 | - | 2 | 118 | |||
FQ219218.1 | - | 2 | 123 | |||
FQ234202.1 | - | 2 | 181 |