Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcnkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173103.1 | NP_775126.1 | 13 | 1354 | ||
XM_006239320.2 | XP_006239382.1 | 16 | 4226 | |||
XM_008764293.1 | XP_008762515.1 | 15 | 3894 | |||
XM_008764294.1 | XP_008762516.1 | 15 | 2160 | |||
XM_008764295.1 | XP_008762517.1 | 14 | 2305 | |||
XM_008764297.1 | XP_008762519.1 | 13 | 2165 | |||
GenBank mRNA | BC081698.1 | - | 13 | 1849 | ||
D26111.1 | - | 14 | 1408 | |||
DQ437671.1 | - | 14 | 1458 | |||
DQ437672.1 | - | 14 | 1351 | |||
DQ437673.1 | - | 13 | 1329 | |||
DQ437674.1 | - | 13 | 1293 | |||
DQ437675.1 | - | 14 | 1301 | |||
DQ437676.1 | - | 13 | 1279 | |||
Z30663.1 | - | 13 | 1354 |