Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173109.1 | NP_775132.1 | 2, 3 | 260 | ||
XM_008762473.1 | XP_008760695.1 | 2, 3 | 265 | |||
GenBank mRNA | AJ515239.1 | - | 2, 3 | 260 |