Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173127.1 | NP_775150.1 | 3, 4 | 466 | ||
GenBank mRNA | FQ226997.1 | - | 3, 4 | 497 | ||
FQ230379.1 | - | 3, 4 | 497 | |||
FQ235277.1 | - | 3, 4 | 497 | |||
M16624.1 | - | 3, 4 | 466 |