Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173128.1 | NP_775151.1 | 1 | 336 | ||
GenBank mRNA | M63665.1 | - | 1 | 336 |