Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103693145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766412.1 | XP_008764634.1 | 2 | 1257 | ||
XM_008766413.1 | XP_008764635.1 | 2 | 1319 | |||
XM_008766414.1 | XP_008764636.1 | 2 | 730 | |||
XM_008766415.1 | XP_008764637.1 | 2 | 1137 |
Gene Symbol: Unc13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173146.2 | NP_775169.4 | 374 | |||
GenBank mRNA | U75361.1 | - | 388 |