Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173147.1 | NP_775170.1 | 4, 5 | 472 | ||
XM_006251537.2 | XP_006251599.1 | 4, 5 | 651 | |||
XM_008770419.1 | XP_008768641.1 | 4, 5 | 651 | |||
XM_008770421.1 | XP_008768643.1 | 4, 5 | 615 | |||
GenBank mRNA | AJ010392.3 | - | 4, 5 | 472 |