Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173320.1 | NP_775442.1 | 3 | 941 | ||
XM_006254377.2 | XP_006254439.1 | 3 | 1055 | |||
GenBank mRNA | AF016185.1 | - | 3 | 941 |
Gene Symbol: LOC286980 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF016186.1 | - | 1 | 435 |
Set Membership: |
G-protein coupled receptor |