Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nampt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177928.3 | NP_808789.1 | 6 | 684 | ||
GenBank mRNA | AB081730.1 | - | 6 | 659 | ||
AY831728.1 | - | 6 | 657 | |||
BC085681.1 | - | 6 | 684 | 7192109 |