Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp27a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006245175.1 | XP_006245237.1 | 2 | 234 | ||
GenBank mRNA | BC061848.1 | - | 2 | 398 | 5621918 | |
M38566.1 | - | 2 | 399 |