Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syngap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113409.2 | NP_001106880.2 | 4 | 540 | ||
NM_181092.4 | NP_851606.3 | 6 | 603 | |||
GenBank mRNA | AF048976.1 | - | 6 | 611 | ||
AF050183.2 | - | 2 | 292 | |||
AF053938.1 | - | 4 | 540 | |||
AF058789.2 | - | 6 | 490 | |||
AF058790.1 | - | 4 | 358 |