Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181626.3 | NP_853657.1 | 3 | 254 | ||
GenBank mRNA | AF508158.1 | - | 3 | 220 | ||
BC070929.1 | - | 3 | 254 | 7096424 | ||
BC078677.1 | - | 3 | 264 | 7133788 |