Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181631.2 | NP_853662.1 | 1 | 108 | ||
XM_008764477.1 | XP_008762699.1 | 1 | 1885 | |||
GenBank mRNA | AY274810.1 | - | 1 | 108 |
Gene Symbol: LOC103689957 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239732.2 | XP_006239794.1 | 1 | 1892 | ||
XM_006239733.2 | XP_006239795.1 | 1 | 1892 |