Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181633.2 | NP_853664.1 | 1 | 167 | ||
GenBank mRNA | AJ564168.1 | - | 1 | 167 |
Set Membership: |
G-protein coupled receptor |