Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021679.1 | NP_067711.1 | 2 | 664 | ||
GenBank mRNA | AF042713.1 | - | 2 | 664 | ||
BC166603.1 | - | 586 | 7936112 |