Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021742.1 | NP_068510.1 | 5 | 517 | ||
XM_006249928.2 | XP_006249990.1 | 4 | 625 | |||
XM_006249929.2 | XP_006249991.1 | 3 | 365 | |||
GenBank mRNA | AB012960.1 | - | 5 | 517 | ||
AB012961.1 | - | 4 | 454 | |||
U47280.1 | - | 2 | 171 |