Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021746.1 | NP_068514.1 | 4 | 551 | ||
XM_008765781.1 | XP_008764003.1 | 4 | 280 | |||
GenBank mRNA | BC081950.1 | - | 4 | 530 | ||
X96488.1 | - | 4 | 551 |