Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002016.2 | NP_001002016.2 | 4 | 997 | ||
GenBank mRNA | BC062018.1 | - | 3 | 552 | 5621637 | |
DN934590.1 | - | 4 | 737 | |||
X66870.1 | - | 4 | 956 | |||
X76297.1 | - | 4 | 719 | |||
X99257.1 | - | 4 | 456 |