Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021859.2 | NP_068631.1 | 3 | 185 | ||
XM_006240989.2 | XP_006241051.1 | 3 | 551 | |||
XM_006240990.2 | XP_006241052.1 | 3 | 211 | |||
XM_008765128.1 | XP_008763350.1 | 3 | 515 | |||
GenBank mRNA | BC087726.1 | - | 3 | 185 | 7320205 |