Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022259.1 | NP_071595.1 | 6 | 1116 | ||
GenBank mRNA | AF156988.1 | - | 6 | 1045 | ||
AF156989.1 | - | 6 | 1116 |