Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022265.2 | NP_071601.2 | 2 | 286 | ||
XM_006231613.2 | XP_006231675.1 | 3 | 567 | |||
XM_006231614.2 | XP_006231676.1 | 3 | 619 | |||
XM_006231615.2 | XP_006231677.1 | 3 | 465 | |||
XM_006231616.2 | XP_006231678.1 | 3 | 442 | |||
GenBank mRNA | AF239739.2 | - | 2 | 286 | ||
BC167751.1 | - | 3 | 474 | 9027822 |