Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmur2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022275.3 | NP_071611.3 | 1 | 658 | ||
GenBank mRNA | AB041229.1 | - | 1 | 658 | ||
AF242875.1 | - | 1 | 532 |
Set Membership: |
G-protein coupled receptor |