Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007361.3 | NP_031387.3 | 2 | 511 | ||
XM_005267405.4 | XP_005267462.1 | 2 | 514 | |||
XM_005267406.4 | XP_005267463.1 | 2 | 514 | |||
XM_005267407.4 | XP_005267464.1 | 2 | 514 | |||
GenBank mRNA | AJ223500.1 | - | 2 | 277 | ||
AK291652.1 | - | 2 | 339 | |||
AK300462.1 | - | 3 | 149 | |||
BC035608.1 | - | 2 | 331 | 5205293 | ||
BX648241.1 | - | 3 | 284 | |||
D86425.1 | - | 2 | 279 |