Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022515.1 | NP_071960.1 | 3 | 159 | ||
GenBank mRNA | BC058473.1 | - | 3 | 184 | 6918141 | |
FQ209984.1 | - | 3 | 178 | |||
FQ220432.1 | - | 3 | 188 | |||
FQ221142.1 | - | 3 | 177 | |||
FQ221221.1 | - | 3 | 184 | |||
FQ221486.1 | - | 3 | 180 | |||
FQ221683.1 | - | 3 | 171 | |||
FQ221688.1 | - | 3 | 180 | |||
FQ221810.1 | - | 3 | 180 | |||
FQ221946.1 | - | 3 | 180 | |||
FQ221988.1 | - | 3 | 184 | |||
FQ222823.1 | - | 3 | 180 | |||
FQ223282.1 | - | 3 | 180 | |||
FQ224199.1 | - | 3 | 180 | |||
FQ224575.1 | - | 3 | 184 | |||
FQ224676.1 | - | 3 | 180 | |||
X78443.1 | - | 3 | 159 |