Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022515.1 | NP_071960.1 | 4 | 309 | ||
GenBank mRNA | BC058473.1 | - | 4 | 334 | 6918141 | |
FQ209984.1 | - | 4 | 328 | |||
FQ220432.1 | - | 4 | 338 | |||
FQ221142.1 | - | 4 | 327 | |||
FQ221221.1 | - | 4 | 334 | |||
FQ221486.1 | - | 4 | 330 | |||
FQ221683.1 | - | 4 | 321 | |||
FQ221688.1 | - | 4 | 330 | |||
FQ221810.1 | - | 4 | 330 | |||
FQ221946.1 | - | 4 | 330 | |||
FQ221988.1 | - | 4 | 334 | |||
FQ222823.1 | - | 4 | 330 | |||
FQ223282.1 | - | 4 | 330 | |||
FQ224199.1 | - | 4 | 330 | |||
FQ224575.1 | - | 4 | 334 | |||
FQ224676.1 | - | 4 | 330 | |||
X78443.1 | - | 4 | 309 |