Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PADI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007365.2 | NP_031391.2 | 5, 6 | 603 | ||
GenBank mRNA | AB023211.1 | - | 5, 6 | 585 | ||
AB030176.1 | - | 5, 6 | 585 | |||
BC009701.1 | - | 5, 6 | 585 | 3901790 |