Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022701.2 | NP_073192.2 | 7 | 689 | ||
XM_006256024.2 | XP_006256086.1 | 7 | 708 | |||
GenBank mRNA | BC064652.1 | - | 7 | 688 | 5598180 | |
CK359020.1 | - | 7 | 689 | |||
DY569467.1 | - | 4 | 371 | |||
U60976.1 | - | 7 | 619 | |||
U60977.1 | - | 6 | 693 |