Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278463.1 | NP_001265392.1 | 4, 5 | 530 | ||
NM_001278464.1 | NP_001265393.1 | 5, 6 | 569 | |||
NM_001278465.1 | NP_001265394.1 | 5, 6 | 569 | |||
NM_005690.4 | NP_005681.2 | 4, 5 | 530 | |||
NM_012062.4 | NP_036192.2 | 4, 5 | 530 | |||
NM_012063.3 | NP_036193.2 | 4, 5 | 530 | |||
XM_005253282.4 | XP_005253339.1 | 5, 6 | 541 | |||
XM_011520543.2 | XP_011518845.1 | 5, 6 | 541 | |||
XM_011520544.2 | XP_011518846.1 | 4, 5 | 410 | |||
XM_017018663.1 | XP_016874152.1 | 4, 5 | 410 | |||
XM_017018664.1 | XP_016874153.1 | 4, 5 | 410 | |||
XM_017018665.1 | XP_016874154.1 | 4, 5 | 410 | |||
GenBank mRNA | AB006965.1 | - | 4, 5 | 401 | ||
AB209070.1 | - | 5, 6 | 446 | |||
AF000430.1 | - | 4, 5 | 438 | |||
AF061795.1 | - | 4, 5 | 387 | |||
AF151685.1 | - | 4, 5 | 387 | |||
AK291094.1 | - | 4, 5 | 410 | |||
AK298568.1 | - | 4, 5 | 410 | |||
AK299926.1 | - | 5, 6 | 482 | |||
AK302565.1 | - | 4, 5 | 530 | |||
AK311606.1 | - | 4, 5 | 525 | |||
BC024590.1 | - | 4, 5 | 400 | 3882922 | ||
BC107763.1 | - | 4, 5 | 414 |