Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012064.3 | NP_036196.1 | 4 | 857 | ||
XM_011538354.1 | XP_011536656.1 | 6 | 674 | |||
XM_017019306.1 | XP_016874795.1 | 4 | 598 | |||
GenBank mRNA | BC074913.2 | - | 4 | 848 | 30915524 | |
BC117474.1 | - | 4 | 857 | 40126025 | ||
BM705396.1 | - | 1 | 81 | |||
KT966395.1 | - | 4 | 806 |