Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR7A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012067.2 | NP_036199.2 | 3, 4 | 592 | ||
XM_011541046.2 | XP_011539348.1 | 3, 4 | 1168 | |||
XM_017000714.1 | XP_016856203.1 | 3, 4 | 326 | |||
GenBank mRNA | AF040639.1 | - | 3, 4 | 599 | ||
AJ271799.1 | - | 3, 4 | 522 | |||
BC025709.1 | - | 3, 4 | 578 | 5224626 | ||
BC031562.1 | - | 3, 4 | 534 | 5184829 | ||
BC042420.1 | - | 3, 4 | 558 | 5186889 | ||
BM920315.1 | - | 3, 4 | 556 |