Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306153.1 | NP_001293082.1 | 2 | 154 | ||
NM_001306154.1 | NP_001293083.1 | 2 | 554 | |||
NM_001306155.1 | NP_001293084.1 | 2 | 386 | |||
NM_001306156.1 | NP_001293085.1 | 2 | 386 | |||
NM_012073.4 | NP_036205.1 | 2 | 554 | |||
GenBank mRNA | AK289353.1 | - | 2 | 152 | ||
AK290393.1 | - | 2 | 195 | |||
AK293341.1 | - | 2 | 223 | |||
AK293437.1 | - | 2 | 157 | |||
AK301760.1 | - | 2 | 146 | |||
AK301985.1 | - | 2 | 193 | |||
AK302368.1 | - | 2 | 259 | |||
AK302383.1 | - | 2 | 384 | |||
AK303086.1 | - | 2 | 183 | |||
AK316376.1 | - | 2 | 141 | |||
BC002971.1 | - | 2 | 129 | |||
BC006543.2 | - | 2 | 169 | 2824199 | ||
BC035499.2 | - | 2 | 216 | 4329194 | ||
BC040027.1 | - | 2 | 186 | |||
BU858349.1 | - | 2 | 322 | |||
D43950.2 | - | 2 | 171 | |||
DB449481.1 | - | 2 | 387 | |||
DC395133.1 | - | 2 | 146 | |||
FJ224303.1 | - | 2 | 305 |