Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023974.1 | NP_076464.1 | 5 | 1335 | ||
XM_008770570.1 | XP_008768792.1 | 5 | 1547 | |||
GenBank mRNA | AF306459.1 | - | 5 | 1335 |