Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280542.1 | NP_001267471.1 | 4 | 441 | ||
NM_001280543.1 | NP_001267472.1 | 5 | 585 | |||
NM_001280544.1 | NP_001267473.1 | 4 | 884 | |||
NM_012074.4 | NP_036206.3 | 4 | 441 | |||
XM_017021670.1 | XP_016877159.1 | 4 | 441 | |||
XM_017021671.1 | XP_016877160.1 | 4 | 1191 | |||
XM_017021672.1 | XP_016877161.1 | 4 | 1193 | |||
XM_017021673.1 | XP_016877162.1 | 4 | 781 | |||
XM_017021674.1 | XP_016877163.1 | 4 | 970 | |||
XM_017021675.1 | XP_016877164.1 | 4 | 1194 | |||
GenBank mRNA | AK024141.1 | - | 4 | 426 | ||
AK126933.1 | - | 4 | 884 | |||
AK294425.1 | - | 5 | 570 | |||
AY803021.1 | - | 4 | 413 | |||
BC026305.1 | - | 4 | 426 | |||
BC060801.1 | - | 4 | 426 | 5260159 | ||
DA010060.1 | - | 4 | 427 | |||
DA407279.1 | - | 4 | 463 | |||
DB140241.1 | - | 4 | 441 |