Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mepe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024142.1 | NP_077056.1 | 3 | 156 | ||
XM_006250634.2 | XP_006250696.1 | 2 | 224 | |||
XM_006250635.2 | XP_006250697.1 | 2 | 1790 | |||
GenBank mRNA | AF260922.1 | - | 3 | 156 | ||
AF530558.1 | - | 3 | 153 | |||
AF530559.1 | - | 2 | 132 |