Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024378.1 | NP_077354.1 | 6 | 1034 | ||
GenBank mRNA | U08255.1 | - | 6 | 1034 | ||
Z17238.1 | - | 4 | 667 |