Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024381.2 | NP_077357.2 | 4 | 391 | ||
XM_006256987.2 | XP_006257049.1 | 4 | 425 | |||
XM_006256988.2 | XP_006257050.1 | 4 | 425 | |||
XM_008773274.1 | XP_008771496.1 | 4 | 406 | |||
XM_008773275.1 | XP_008771497.1 | 4 | 406 | |||
XM_008773276.1 | XP_008771498.1 | 4 | 406 | |||
XM_008773278.1 | XP_008771500.1 | 4 | 405 | |||
GenBank mRNA | D16102.1 | - | 4 | 394 |