Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277073.1 | NP_001264002.1 | 3, 4 | 340 | ||
NM_024387.2 | NP_077363.1 | 3, 4 | 384 | |||
XM_006245827.2 | XP_006245889.1 | 3, 4 | 562 | |||
XM_008767500.1 | XP_008765722.1 | 2, 3 | 1291 | |||
GenBank mRNA | BC062061.1 | - | 3, 4 | 299 | 5599345 | |
FM134576.1 | - | 3, 4 | 277 | |||
FQ225862.1 | - | 3, 4 | 337 | |||
J05405.1 | - | 3, 4 | 376 |
Gene Symbol: Hmox2-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF058787.1 | - | 1258 |