Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifitm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030833.1 | NP_110460.1 | 2 | 562 | ||
GenBank mRNA | AF164040.1 | - | 2 | 562 | ||
BC060563.1 | - | 529 | 6918334 |