Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030839.3 | NP_110466.2 | 5 | 959 | ||
XM_008762036.1 | XP_008760258.1 | 5 | 1553 | |||
GenBank mRNA | AF313453.1 | - | 5 | 1144 | ||
AF375466.1 | - | 5 | 959 |