Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030872.1 | NP_110499.1 | 3, 4 | 435 | ||
GenBank mRNA | AF237719.1 | - | 3, 4 | 435 | ||
BC061823.1 | - | 3, 4 | 399 | 5599188 | ||
U10357.1 | - | 3, 4 | 435 |