Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031014.2 | NP_112276.2 | 4, 5 | 352 | ||
GenBank mRNA | BC072496.1 | - | 4, 5 | 352 | 7096880 | |
D89514.1 | - | 4, 5 | 345 |