Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031024.1 | NP_112286.1 | 5 | 400 | ||
XM_006253655.2 | XP_006253717.1 | 5 | 486 | |||
XM_006253656.2 | XP_006253718.1 | 5 | 485 | |||
XM_006253657.2 | XP_006253719.1 | 5 | 484 | |||
XM_008771507.1 | XP_008769729.1 | 5 | 1090 | |||
GenBank mRNA | AB015042.1 | - | 5 | 347 | ||
AB514558.1 | - | 5 | 347 | |||
BC139847.1 | - | 5 | 402 | 8368018 | ||
FQ217148.1 | - | 5 | 429 | |||
X59267.1 | - | 5 | 400 |