Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gna12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031034.2 | NP_112296.1 | 2 | 407 | ||
GenBank mRNA | D85760.1 | - | 2 | 441 | ||
DQ120479.1 | - | 2 | 376 | |||
DQ120480.1 | - | 2 | 376 |