Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031078.2 | NP_112340.1 | 16 | 2211 | ||
XM_008762922.1 | XP_008761144.1 | 13 | 1714 | |||
XM_008762923.1 | XP_008761145.1 | 12 | 1442 | |||
XM_008762924.1 | XP_008761146.1 | 12 | 1442 | |||
GenBank mRNA | L41045.1 | - | 16 | 2211 |