Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031101.1 | NP_112363.1 | 4 | 352 | ||
GenBank mRNA | BC058143.1 | - | 4 | 355 | 6919851 | |
BC086577.1 | - | 2 | 576 | |||
FQ221209.1 | - | 4 | 373 | |||
FQ222628.1 | - | 4 | 350 | |||
FQ222810.1 | - | 4 | 350 | |||
FQ223361.1 | - | 4 | 371 | |||
FQ224984.1 | - | 4 | 348 | |||
FQ225827.1 | - | 2 | 118 | |||
FQ226346.1 | - | 4 | 349 | |||
FQ228254.1 | - | 4 | 348 | |||
FQ229622.1 | - | 4 | 350 | |||
X78327.1 | - | 4 | 352 |
Gene Symbol: LOC100359922 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765903.1 | XP_008764125.1 | 1 | 349 |