Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031104.1 | NP_112366.1 | 4 | 165 | ||
GenBank mRNA | BC058466.1 | - | 3 | 181 | 6887477 | |
BC082750.1 | - | 3 | 156 | 7300771 | ||
FQ222147.1 | - | 3 | 174 | |||
FQ222660.1 | - | 3 | 174 | |||
FQ222798.1 | - | 3 | 175 | |||
FQ224693.1 | - | 3 | 173 | |||
FQ228373.1 | - | 3 | 171 | |||
FQ228982.1 | - | 3 | 173 | |||
X78444.1 | - | 4 | 165 |
Gene Symbol: LOC100360057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729392.4 | XP_002729438.2 | 1 | 178 |