Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031112.1 | NP_112374.1 | 4, 5 | 430 | ||
GenBank mRNA | FQ209499.1 | - | 2, 3 | 204 | ||
FQ221116.1 | - | 2, 3 | 204 | |||
FQ221331.1 | - | 4, 5 | 435 | |||
FQ221519.1 | - | 2, 3 | 204 | |||
FQ221824.1 | - | 2, 3 | 204 | |||
FQ222238.1 | - | 4, 5 | 435 | |||
FQ223039.1 | - | 4, 5 | 435 | |||
FQ223185.1 | - | 2, 3 | 215 | |||
FQ225772.1 | - | 4, 5 | 435 | |||
FQ228365.1 | - | 2, 3 | 204 | |||
FQ228683.1 | - | 2, 3 | 204 | |||
FQ229436.1 | - | 2, 3 | 204 | |||
X51538.1 | - | 4, 5 | 430 | |||
X52445.1 | - | 4, 5 | 430 |